Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Pyridyl)thiazolidine, 97%

Chemical Identifiers

Chemical Name or Material 2-(3-Pyridyl)thiazolidine
CAS 700-93-6
Formula Weight 166.24
MDL Number MFCD01691463
Synonym 3-(2-Thiazolidinyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6207903

 
alfa aesar™
H6207903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6207906

 
alfa aesar™
H6207906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62079MD

 
alfa aesar™
H62079MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.24
Chemical Name or Material 2-(3-Pyridyl)thiazolidine
Physical Form Solid
Assay 97%
MDL Number MFCD01691463
CAS 700-93-6
Synonym 3-(2-Thiazolidinyl)pyridine
RTECS Number UT8571100
Quantity 1g