missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Pyridyl)thiazolidine, 97%

Manufacturer:  Alfa Aesar™ H6207906

 View more versions of this product

Catalog No. AAH6207906



Specifications

Specifications

2-(3-Pyridyl)thiazolidine
700-93-6
MFCD01691463
SMYGUEYUDFIZCL-UHFFFAOYSA-N
2-pyridin-3-yl-1,3-thiazolidine
120769
Solid
5g
C8H10N2S
2-3-pyridyl thiazolidine
C1CSC(N1)C2=CN=CC=C2
166.242
166.24
97%

Chemical Identifiers

700-93-6
166.242
SMYGUEYUDFIZCL-UHFFFAOYSA-N
120769
C1CSC(N1)C2=CN=CC=C2
C8H10N2S
MFCD01691463
2-3-pyridyl thiazolidine
2-pyridin-3-yl-1,3-thiazolidine
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : UT8571100

TSCA : No

Recommended Storage : Ambient temperatures

Documents