Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Pyridyl)thiazolidine-4-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 2-(3-Pyridyl)thiazolidine-4-carboxylic acid
CAS 59777-95-6
Formula Weight 210.25
MDL Number MFCD01115161
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6221403

  SDS
alfa aesar™
H6221403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62214MD

  SDS
alfa aesar™
H62214MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6221406

  SDS
alfa aesar™
H6221406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.25
Assay 97%
Physical Form Solid
Quantity 1g
MDL Number MFCD01115161
Chemical Name or Material 2-(3-Pyridyl)thiazolidine-4-carboxylic acid
CAS 59777-95-6