missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Pyridyl)thiazolidine-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H6221406

 View more versions of this product

Catalog No. AAH6221406



Specifications

Specifications

2-(3-Pyridyl)thiazolidine-4-carboxylic acid
59777-95-6
MFCD01115161
FSNGLHIMQHWTNF-UHFFFAOYSA-N
2-pyridin-3-yl-1,3-thiazolidine-4-carboxylic acid
327593
Solid
5g
C9H10N2O2S
2-3-piridyl thiazolidine-4-carboxylic acid
C1C(NC(S1)C2=CN=CC=C2)C(=O)O
210.251
210.25
97%

Chemical Identifiers

59777-95-6
210.251
FSNGLHIMQHWTNF-UHFFFAOYSA-N
327593
C1C(NC(S1)C2=CN=CC=C2)C(=O)O
C9H10N2O2S
MFCD01115161
2-3-piridyl thiazolidine-4-carboxylic acid
2-pyridin-3-yl-1,3-thiazolidine-4-carboxylic acid
Documents