missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(3-Pyridyl)thiazole-4-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5183806
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2-(3-Pyridyl)thiazole-4-carboxylic acid is a pyridylthiazole compound that has use in a variety of diverse applications from highly luminescent heterocyclic compounds to potential antiulcer drugs, to the formation of double and triple helicates with Cu(II) and to the mesoscopically ordered pH-responsive hybrid materials.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2-(3-Pyridyl)thiazole-4-carboxylic acid | |
C9H6N2O2S | |
5 g | |
FOQFGMAZUTUELM-UHFFFAOYSA-N | |
2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid | |
736513 | |
97% |
39067-29-3 | |
MFCD00052304 | |
2-pyridin-3-yl thiazole-4-carboxylic acid, 2-3-pyridyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-3-pyridinyl, 2-pyridin-3-yl-thiazole-4-carboxylic acid, 2-3-pyridyl thiazole-4-carboxylic acid, 2-3-pyridinyl-4-thiazolecarboxylic acid, 2-pyrid-3-yl thiazole-4-carboxylic acid, 2-3-pyridinyl-1,3-thiazole-4-carboxylic acid | |
C1=CC(=CN=C1)C2=NC(=CS2)C(=O)O | |
206.219 | |
206.22 |
Chemical Identifiers
39067-29-3 | |
206.219 | |
FOQFGMAZUTUELM-UHFFFAOYSA-N | |
736513 | |
C1=CC(=CN=C1)C2=NC(=CS2)C(=O)O |
C9H6N2O2S | |
MFCD00052304 | |
2-pyridin-3-yl thiazole-4-carboxylic acid, 2-3-pyridyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-3-pyridinyl, 2-pyridin-3-yl-thiazole-4-carboxylic acid, 2-3-pyridyl thiazole-4-carboxylic acid, 2-3-pyridinyl-4-thiazolecarboxylic acid, 2-pyrid-3-yl thiazole-4-carboxylic acid, 2-3-pyridinyl-1,3-thiazole-4-carboxylic acid | |
2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures