Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Pyridyl)benzimidazole, 97%

$251.64 - $251.64

Chemical Identifiers

Chemical Name or Material 2-(3-Pyridyl)benzimidazole
CAS 1137-67-3
Molecular Formula C12H9N3
Formula Weight 195.22
MDL Number MFCD00022665
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5178514

  SDS
alfa aesar™
H5178514
25g Each for $251.64
Description & Specifications

Specifications

Formula Weight 195.22
Melting Point 254° to 260°C
Molecular Formula C12H9N3
CAS 1137-67-3
Health Hazard 1 Warning
Chemical Name or Material 2-(3-Pyridyl)benzimidazole
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Odor Odorless
Quantity 25g
MDL Number MFCD00022665
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.