missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Pyridyl)benzimidazole-6-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5459303

 View more versions of this product

Catalog No. AAH5459303



Specifications

Specifications

2-(3-Pyridyl)benzimidazole-6-carboxylic acid
312496-16-5
MFCD01414693
DHRPCXYIVJXXJR-UHFFFAOYSA-N
2-pyridin-3-yl-3H-benzimidazole-5-carboxylic acid
2771805
97%
1g
C13H9N3O2
1h-benzimidazole-6-carboxylicacid, 2-3-pyridinyl
C1=CC(=CN=C1)C2=NC3=C(N2)C=C(C=C3)C(=O)O
239.234
239.23

Chemical Identifiers

312496-16-5
239.234
DHRPCXYIVJXXJR-UHFFFAOYSA-N
2771805
C1=CC(=CN=C1)C2=NC3=C(N2)C=C(C=C3)C(=O)O
C13H9N3O2
MFCD01414693
1h-benzimidazole-6-carboxylicacid, 2-3-pyridinyl
2-pyridin-3-yl-3H-benzimidazole-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents