missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione, 97%, Alfa Aesar™

Chemical Identifiers

CAS 942119-32-6
Molecular Formula C20H11N3O2
Molecular Weight (g/mol) 325.327
MDL Number MFCD20265346
InChI Key IOBRUMYBBWCOCR-UHFFFAOYSA-N
Synonym 2-3-pyridyl-1h-anthra 1,2-d imidazole-6,11-dione
PubChem CID 16659185
IUPAC Name 2-pyridin-3-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CN=CC=C5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54812MD View Documents Thermo Fisher Scientific
H54812MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5481203 View Documents Thermo Fisher Scientific
H5481203
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5481206 View Documents Thermo Fisher Scientific
H5481206
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

942119-32-6
325.327
IOBRUMYBBWCOCR-UHFFFAOYSA-N
16659185
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CN=CC=C5
C20H11N3O2
MFCD20265346
2-3-pyridyl-1h-anthra 1,2-d imidazole-6,11-dione
2-pyridin-3-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione

Specifications

250mg
C20H11N3O2
2-3-pyridyl-1h-anthra 1,2-d imidazole-6,11-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CN=CC=C5
325.327
325.33
2-(3-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione
942119-32-6
MFCD20265346
IOBRUMYBBWCOCR-UHFFFAOYSA-N
2-pyridin-3-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
16659185
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures