missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5481203

 View more versions of this product

Catalog No. AAH5481203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(3-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione
942119-32-6
MFCD20265346
IOBRUMYBBWCOCR-UHFFFAOYSA-N
2-pyridin-3-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
16659185
97%
1g
C20H11N3O2
2-3-pyridyl-1h-anthra 1,2-d imidazole-6,11-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CN=CC=C5
325.327
325.33

Chemical Identifiers

942119-32-6
325.327
IOBRUMYBBWCOCR-UHFFFAOYSA-N
16659185
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CN=CC=C5
C20H11N3O2
MFCD20265346
2-3-pyridyl-1h-anthra 1,2-d imidazole-6,11-dione
2-pyridin-3-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents