Learn More
2,3-Pyridinedicarboxylic acid, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131855000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 3-Pyridinedicarboxylic acid | |
89-00-9 | |
100.0 | |
99% | |
C7H5NO4 | |
500g | |
15, 8190 | |
Solubility in water: 0.55%. Other solubilities: soluble in alkalies,slightly soluble in alcohol,practically insoluble in ether and benzene | |
C1=CC(=C(N=C1)C(=O)O)C(=O)O | |
167.12 | |
CHEBI:16675 | |
99% |
99% | |
98.5 | |
Authentic | |
Plastic bottle | |
MFCD00006295 | |
22, 150 | |
quinolinic acid, 2,3-pyridinedicarboxylic acid, quinolinate, pyridine-2,3-dicarboxylate, unii-f6f0hk1urn, 2,3-pyridinedicarboxylicacid, pyridine-2,3-carboxylate, f6f0hk1urn, pyridin-2,3-dicarbonsaeure, 2,3-pyridinedicarboxylate | |
GJAWHXHKYYXBSV-UHFFFAOYSA-N | |
pyridine-2,3-dicarboxylic acid | |
1066 | |
167.12 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 201-874-8
RTECSNumber : US7967250
TSCA : TSCA
RUO â Research Use Only