Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Piperidinyloxy)benzonitrile, 99%

$371.81 - $371.81

Chemical Identifiers

Chemical Name or Material 2-(3-Piperidinyloxy)benzonitrile
CAS 902836-51-5
Molecular Formula C12H14N2O
Formula Weight 202.26g/mol
MDL Number MFCD08061059
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5091803

 
alfa aesar™
H50918-03
Each for $371.81
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 2-(3-Piperidinyloxy)benzonitrile
CAS 902836-51-5
Assay 99%
Molecular Formula C12H14N2O
Formula Weight 202.26g/mol
MDL Number MFCD08061059
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.