Please login to your online account to display your discounted pricing

2-(3-Phenylpropyl)pyridine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC381940050

 View more versions of this product

Catalog No. AC381940050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 197.28
Quantity 5g
Color Clear yellow
Density 1.0150g/mL
CAS Max % 100.0
Chemical Name or Material 2-(3-Phenylpropyl)pyridine
Assay Percent Range 98%
CAS 2110-18-1
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Molecular Formula C14H15N
MDL Number MFCD00191237
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 2-(3-Phenylpropyl)pyridine
CAS 2110-18-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H15N
Formula Weight 197.28
MDL Number MFCD00191237
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents