Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Pentanedione, 97%

Chemical Identifiers

Chemical Name or Material 2,3-Pentanedione
CAS 600-14-6
Molecular Formula C5H8O2
Formula Weight 100.12
MDL Number MFCD00009313
UN Number UN1224
Beilstein 1699638
Synonym Acetylpropionyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1311614

  SDS
alfa aesar™
A1311614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1311636

  SDS
alfa aesar™
A1311636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.12
Chemical Name or Material 2,3-Pentanedione
MDL Number MFCD00009313
UN Number UN1224
Beilstein 1699638
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number SA1850000
Flash Point 18°C (64°F)
Odor Odorless
Quantity 25g
TSCA TSCA
Assay 97%
Molecular Formula C5H8O2
CAS 600-14-6
Synonym Acetylpropionyl
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Boiling Point 110° to 112°C
Melting Point −52°C
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C