missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2',3'-O-Isopropylideneuridine, 99%, ACROS Organics™
Chemical Identifiers
CAS | 362-43-6 |
---|---|
Molecular Formula | C12H16N2O6 |
Molecular Weight (g/mol) | 284.268 |
MDL Number | MFCD00034509 |
InChI Key | GFDUSNQQMOENLR-PEBGCTIMSA-N |
Synonym | 1-1r,2r,4r,5r-4-hydroxymethyl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oct-2-yl-1,3-dihydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-3h-pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4 1h,3h-dione, 2',3'-isopropylideneuridine, 2',3'-o-isopropylidene uridine, 2',3'-o-isopropylideneuridine, 2',3'-o-isopropylideneuridine hplc, 2/',3/'-isopropylideneuridine |
PubChem CID | 64967 |
IUPAC Name | 1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]pyrimidine-2,4-dione |
SMILES | CC1(OC2C(OC(C2O1)N3C=CC(=O)NC3=O)CO)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC122740010
|
Thermo Fisher Scientific
122740010 |
1g | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
362-43-6 | |
284.268 | |
GFDUSNQQMOENLR-PEBGCTIMSA-N | |
64967 | |
CC1(OC2C(OC(C2O1)N3C=CC(=O)NC3=O)CO)C |
C12H16N2O6 | |
MFCD00034509 | |
1-1r,2r,4r,5r-4-hydroxymethyl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oct-2-yl-1,3-dihydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-3h-pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4 1h,3h-dione, 2',3'-isopropylideneuridine, 2',3'-o-isopropylidene uridine, 2',3'-o-isopropylideneuridine, 2',3'-o-isopropylideneuridine hplc, 2/',3/'-isopropylideneuridine | |
1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]pyrimidine-2,4-dione |
Specifications
Authentic | |
White | |
1g | |
98.5 | |
99% | |
MFCD00034509 | |
GFDUSNQQMOENLR-PEBGCTIMSA-N | |
1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]pyrimidine-2,4-dione | |
64967 | |
Fine Crystalline Powder or Needles | |
2', 3'-O-Isopropylideneuridine |
Glass Bottle | |
163.0°C to 166.0°C | |
362-43-6 | |
100.0 | |
C12H16N2O6 | |
1-1r,2r,4r,5r-4-hydroxymethyl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oct-2-yl-1,3-dihydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-3h-pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4 1h,3h-dione, 2',3'-isopropylideneuridine, 2',3'-o-isopropylidene uridine, 2',3'-o-isopropylideneuridine, 2',3'-o-isopropylideneuridine hplc, 2/',3/'-isopropylideneuridine | |
CC1(OC2C(OC(C2O1)N3C=CC(=O)NC3=O)CO)C | |
284.268 | |
284.26 | |
99% |