Please login to your online account to display your discounted pricing

2',3'-O-Isopropylideneuridine, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC122740050

 View more versions of this product

Catalog No. AC122740050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 284.26
Assay ≥98.5%
Physical Form Fine Crystalline Powder or Needles
Quantity 5g
Color White
Melting Point 163° to 166°C
CAS Max % 100.0
Chemical Name or Material 2', 3'-O-Isopropylideneuridine
Molecular Formula C12H16N2O6
Assay Percent Range 99%
CAS 362-43-6
CAS Min % 98.5
MDL Number MFCD00034509
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2', 3'-O-Isopropylideneuridine
CAS 362-43-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H16N2O6
Formula Weight 284.26
MDL Number MFCD00034509