missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3'-O-Isopropylideneadenosine, 98%

Manufacturer:  Alfa Aesar™ H5340114

 View more versions of this product

Catalog No. AAH5340114



Description

Description

Applications
2′,3′-O-Isopropylideneadenosine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

2',3'-O-Isopropylideneadenosine
221°C to 222°C
362-75-4
MFCD00005756
2',3'-o-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 2',3'-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl methanol, 2 ,3-o-isopropylideneadenosine, 1r,2r,4r,5r-4-6-aminopurin-9-yl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oc t-2-yl methan-1-ol, 3ar,4r,6r,6ar-4-6-aminopurin-9-yl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-6-yl methanol, 3ar,4r,6r,6ar-6-6-aminopurin-9-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem14185
CC1(OC2C(OC(C2O1)N3C=NC4=C3N=CN=C4N)CO)C
307.31
307.31
−99° (c=1 in Dioxane)
25g
C13H17N5O4
43435
LCCLUOXEZAHUNS-WOUKDFQISA-N
[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol
2723654
98%

Chemical Identifiers

362-75-4
307.31
LCCLUOXEZAHUNS-WOUKDFQISA-N
2723654
CC1(OC2C(OC(C2O1)N3C=NC4=C3N=CN=C4N)CO)C
C13H17N5O4
MFCD00005756
2',3'-o-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 2',3'-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl methanol, 2 ,3-o-isopropylideneadenosine, 1r,2r,4r,5r-4-6-aminopurin-9-yl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oc t-2-yl methan-1-ol, 3ar,4r,6r,6ar-4-6-aminopurin-9-yl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-6-yl methanol, 3ar,4r,6r,6ar-6-6-aminopurin-9-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem14185
[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol
SDS Documents