Please login to your online account to display your discounted pricing

(-)-2,3-O-Isopropylidene-D-threitol, 99%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  221020010

 View more versions of this product

Catalog No. AC221020010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 162.19
Quantity 1g
Color Off-white
Specific Rotation Condition −3.10° (26°C c=5,CHCl3)
EINECS Number 277-391-1
TSCA TSCA
Melting Point 42° to 48°C
Beilstein 19, III, 971
CAS Max % 100.0
Chemical Name or Material (-)-2, 3-O-Isopropylidene-D-threitol
Name Note 99%
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H14O4
MDL Number MFCD00009761
CAS 73346-74-4
Synonym D(-)-2, 2-Dimethyl-1, 3-dioxolane-4
CAS Min % 98.5
Specific Rotation -2.1° to -4.1° (26°C, 589nm) (c=5, CHCl3)

Specifications

Chemical Name or Material (-)-2, 3-O-Isopropylidene-D-threitol
Name Note 99%
CAS 73346-74-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H14O4
Formula Weight 162.19
MDL Number MFCD00009761
Synonym D(-)-2, 2-Dimethyl-1, 3-dioxolane-4