Please login to your online account to display your discounted pricing

(-)-2,3-O-Isopropylidene-D-threitol, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material (-)-2, 3-O-Isopropylidene-D-threitol
Name Note 99%
CAS 73346-74-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H14O4
Formula Weight 162.19
MDL Number MFCD00009761
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC221020010

  SDS
thermo fisher scientific
221020010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 162.19
Quantity 1g
Color Off-white
Specific Rotation Condition −3.10° (26°C c=5,CHCl3)
TSCA TSCA
EINECS Number 277-391-1
Melting Point 42° to 48°C
Beilstein 19, III, 971
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Specific Rotation -2.1° to -4.1° (26°C, 589nm) (c=5, CHCl3)
Synonym D(-)-2, 2-Dimethyl-1, 3-dioxolane-4
Chemical Name or Material (-)-2, 3-O-Isopropylidene-D-threitol
Assay Percent Range 99%
MDL Number MFCD00009761
CAS 73346-74-4
Name Note 99%
Molecular Formula C7H14O4