missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-O-Isopropylidene-D-erythronolactone, 98%

For use in spiroannulated carbohydrate synthesis

Manufacturer:  Alfa Aesar™ J6687103

 View more versions of this product

Catalog No. AAJ6687103



Description

Description

Applications
It undergoes Aldol condensations with silyl ketene acetals.1 Employed in spiroannulated carbohydrate synthesis.2 Convergent syntheses of a hydroxylated indolizidine,3 carbohydrate substituted benzoquinones,4 and of the oxazole segment of calyculin5 have been accomplished using this chiral synthon.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents.
Specifications

Specifications

2,3-O-Isopropylidene-D-erythronolactone
White
1g
C7H10O4
1282952
Soluble in water
CC1(OC2COC(=O)C2O1)C
158.153
158.15
98%
−118° (c=1 in Water)
67°C to 69°C
25581-41-3
MFCD00134440
--2,3-o-isopropylidene-d-erythronolactone, 2,3-o-isopropylidene-d-erythronolactol, 2,3-o-isopropylidene-d-erythronolactone, 2-o,3-o-isopropylidene-d-erythronic acid gamma-lactone, 3ar,6ar-2,2-dimethyl-6,6a-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, 3ar,6ar-2,2-dimethyl-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, 3ar,6ar-2,2-dimethyldihydrofuro 3,4-d 1,3 dioxol-4 3ah-one, a-lactone, d-2,3-o-isopropylidene-erythronic acid, pubchem19725
WHPSMBYLYRPVGU-RFZPGFLSSA-N
(3aR,6aR)-2,2-dimethyl-6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxol-4-one
386843
Powder

Chemical Identifiers

25581-41-3
158.153
WHPSMBYLYRPVGU-RFZPGFLSSA-N
386843
CC1(OC2COC(=O)C2O1)C
C7H10O4
MFCD00134440
--2,3-o-isopropylidene-d-erythronolactone, 2,3-o-isopropylidene-d-erythronolactol, 2,3-o-isopropylidene-d-erythronolactone, 2-o,3-o-isopropylidene-d-erythronic acid gamma-lactone, 3ar,6ar-2,2-dimethyl-6,6a-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, 3ar,6ar-2,2-dimethyl-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, 3ar,6ar-2,2-dimethyldihydrofuro 3,4-d 1,3 dioxol-4 3ah-one, a-lactone, d-2,3-o-isopropylidene-erythronic acid, pubchem19725
(3aR,6aR)-2,2-dimethyl-6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxol-4-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents