Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-O-Isopropylidene-alpha,beta-D-ribofuranose

$105.03 - $418.88

Chemical Identifiers

Chemical Name or Material 2,3-O-Isopropylidene-alpha,beta-D-ribofuranose
CAS 13199-25-2
Molecular Formula C8H14O5
Formula Weight 190.2
MDL Number MFCD01075718
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1598803

  SDS
alfa aesar™
L1598803
1g Each for $105.03

AAL1598806

  SDS
alfa aesar™
L1598806
5g Each for $418.88
Description & Specifications

Specifications

Formula Weight 190.2
Chemical Name or Material 2,3-O-Isopropylidene-alpha,beta-D-ribofuranose
CAS 13199-25-2
MDL Number MFCD01075718
Molecular Formula C8H14O5
Quantity 1g
Optical Rotation −27° (c=1 in Chloroform)
Recommended Storage 2° to 8°C

Applications
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose is used as a pharmaceutical intermediate.

Solubility
Soluble in acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

2° to 8°C