Please login to your online account to display your discounted pricing

(+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane, 98%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  218012500

 View more versions of this product

Catalog No. AC218012500

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 498.53
Physical Form Powder
Assay ≥97.5%
Quantity 250mg
Color White
Merck Index 15, 3319
TSCA TSCA
EINECS Number 253-307-9
Melting Point 87° to 92°C
Specific Rotation Condition + 12.50 (20.00°C c=5,benzene)
Fieser 04,273; 05,360; 06,309; 09,259; 11,277
CAS Max % 100.0
Specific Rotation +25.0° to +27.0° (20°C, 589nm) (c=2, CHCl3)
Health Hazard 1 AIR SENSITIVE
CAS Min % 97.5
Synonym (+)-DIOP
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane
MDL Number MFCD00009760
Assay Percent Range 98%
Molecular Formula C31H32O2P2
CAS 37002-48-5
Name Note 98%

Specifications

Chemical Name or Material (+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane
Name Note 98%
CAS 37002-48-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C31H32O2P2
Formula Weight 498.53
MDL Number MFCD00009760
Synonym (+)-DIOP
Accessories

AIR SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents