Please login to your online account to display your discounted pricing

(+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  218010010

 View more versions of this product

Catalog No. AC218010010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 498.53
Quantity 1g
Color White
EINECS Number 253-307-9
TSCA TSCA
Specific Rotation Condition + 12.50 (20.00°C c=5,benzene)
Melting Point 87° to 92°C
Fieser 04,273; 05,360; 06,309; 09,259; 11,277
Merck Index 15, 3319
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material (+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C31H32O2P2
MDL Number MFCD00009760
Synonym (+)-DIOP
CAS Min % 97.5
Health Hazard 1 AIR SENSITIVE
Specific Rotation +25.0° to +27.0° (20°C, 589nm) (c=2, CHCl3)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 37002-48-5

Specifications

Chemical Name or Material (+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane
Name Note 98%
CAS 37002-48-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C31H32O2P2
Formula Weight 498.53
MDL Number MFCD00009760
Synonym (+)-DIOP
Accessories

AIR SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents