missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-O-Isopropylidene-1-O-methyl-D-ribosic acid, 97%

$41.64 - $116.28

Chemical Identifiers

CAS 54622-95-6
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.205
MDL Number MFCD01632660
InChI Key BTFKDZSYIKUCGF-LZCNBPATSA-N
Synonym 2,3-o-isopropylidene-1-o-methyl-d-ribosic acid, 3as,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxole-4-carboxylic acid, 3as,6r,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxole-4-carboxylic acid
PubChem CID 14188418
IUPAC Name (3aR,4R,6S,6aR)-4-methoxy-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole-6-carboxylic acid
SMILES CC1(OC2C(O1)C(OC2C(=O)O)OC)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1758303 View Documents Alfa Aesar™
L1758303
1g Each for $116.28
 
AAL17583MD View Documents Alfa Aesar™
L17583MD
250mg Each for $41.64
 
Description

Description

Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

54622-95-6
218.205
BTFKDZSYIKUCGF-LZCNBPATSA-N
14188418
CC1(OC2C(O1)C(OC2C(=O)O)OC)C
C9H14O6
MFCD01632660
2,3-o-isopropylidene-1-o-methyl-d-ribosic acid, 3as,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxole-4-carboxylic acid, 3as,6r,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxole-4-carboxylic acid
(3aR,4R,6S,6aR)-4-methoxy-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole-6-carboxylic acid

Specifications

129°C to 130°C
54622-95-6
MFCD01632660
BTFKDZSYIKUCGF-LZCNBPATSA-N
(3aR,4R,6S,6aR)-4-methoxy-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole-6-carboxylic acid
14188418
97%
1g
C9H14O6
2,3-o-isopropylidene-1-o-methyl-d-ribosic acid, 3as,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxole-4-carboxylic acid, 3as,6r,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxole-4-carboxylic acid
CC1(OC2C(O1)C(OC2C(=O)O)OC)C
218.205
218.21
2,3-O-Isopropylidene-1-O-methyl-D-ribosic acid