missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-O-Isopropylidene-1-O-methyl-D-ribosic acid, 97%

Manufacturer:  Alfa Aesar™ L17583MD

 View more versions of this product

Catalog No. AAL17583MD



Description

Description

Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications

Specifications

2,3-O-Isopropylidene-1-O-methyl-D-ribosic acid
250mg
C9H14O6
2,3-o-isopropylidene-1-o-methyl-d-ribosic acid
CC1(OC2C(O1)C(OC2C(=O)O)OC)C
218.205
218.21
129°C to 130°C
54622-95-6
MFCD01632660
BTFKDZSYIKUCGF-LZCNBPATSA-N
(3aR,4R,6S,6aR)-4-methoxy-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole-6-carboxylic acid
14188418
97%

Chemical Identifiers

54622-95-6
218.205
BTFKDZSYIKUCGF-LZCNBPATSA-N
14188418
CC1(OC2C(O1)C(OC2C(=O)O)OC)C
C9H14O6
MFCD01632660
2,3-o-isopropylidene-1-o-methyl-d-ribosic acid
(3aR,4R,6S,6aR)-4-methoxy-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole-6-carboxylic acid
Documents