missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Methylphenyl)benzimidazole, 95%

$115.66 - $1,517.45

Chemical Identifiers

CAS 6528-83-2
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD01220491
InChI Key USLJYASOSQJYGN-UHFFFAOYSA-N
Synonym 2-3-methylphenyl benzimidazole, 2-3-methylphenyl-1h-benzimidazole, 2-m-tolyl-1h-benzo d imidazole, 2-3-methylphenyl-1h-1,3-benzodiazole, 2-3-methylphenyl-1h-benzo d imidazole, 2-m-tolyl-1h-benzoimidazole, cbmicro_005935, cambridge id 5978943, 2-m-tolyl-1h-benzoimidazole, 2-3-methylphenyl-benzimidazole
PubChem CID 2879476
IUPAC Name 2-(3-methylphenyl)-1H-benzimidazole
SMILES CC1=CC=CC(=C1)C2=NC3=CC=CC=C3N2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64264MD View Documents Alfa Aesar™
H64264MD
250mg Each for $115.66
 
AAH6426403 View Documents Alfa Aesar™
H6426403
1g Each for $345.43
 
AAH6426406 View Documents Alfa Aesar™
H6426406
5g Each for $1,517.45
 
Specifications

Chemical Identifiers

6528-83-2
208.264
USLJYASOSQJYGN-UHFFFAOYSA-N
2879476
CC1=CC=CC(=C1)C2=NC3=CC=CC=C3N2
C14H12N2
MFCD01220491
2-3-methylphenyl benzimidazole, 2-3-methylphenyl-1h-benzimidazole, 2-m-tolyl-1h-benzo d imidazole, 2-3-methylphenyl-1h-1,3-benzodiazole, 2-3-methylphenyl-1h-benzo d imidazole, 2-m-tolyl-1h-benzoimidazole, cbmicro_005935, cambridge id 5978943, 2-m-tolyl-1h-benzoimidazole, 2-3-methylphenyl-benzimidazole
2-(3-methylphenyl)-1H-benzimidazole

Specifications

White
6528-83-2
MFCD01220491
USLJYASOSQJYGN-UHFFFAOYSA-N
2-(3-methylphenyl)-1H-benzimidazole
2879476
Solid
2-(3-Methylphenyl)benzimidazole
250mg
C14H12N2
2-3-methylphenyl benzimidazole, 2-3-methylphenyl-1h-benzimidazole, 2-m-tolyl-1h-benzo d imidazole, 2-3-methylphenyl-1h-1,3-benzodiazole, 2-3-methylphenyl-1h-benzo d imidazole, 2-m-tolyl-1h-benzoimidazole, cbmicro_005935, cambridge id 5978943, 2-m-tolyl-1h-benzoimidazole, 2-3-methylphenyl-benzimidazole
CC1=CC=CC(=C1)C2=NC3=CC=CC=C3N2
208.264
208.26
95%