Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Methylphenyl)acetamidoxime, 97%

Chemical Identifiers

Chemical Name or Material 2-(3-Methylphenyl)acetamidoxime
Formula Weight 164.2
Synonym 2-(m-Tolyl)acetamidoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6678403

 
alfa aesar™
H6678403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66784MD

 
alfa aesar™
H66784MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.2
Chemical Name or Material 2-(3-Methylphenyl)acetamidoxime
Assay 97%
Physical Form Crystalline Solid
Synonym 2-(m-Tolyl)acetamidoxime
Color White to pale brown
Quantity 1g