Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Methoxyphenyl)thioacetamide, 97%

$111.65 - $373.68

Chemical Identifiers

Chemical Name or Material 2-(3-Methoxyphenyl)thioacetamide
CAS 35582-11-7
Molecular Formula C9H11NOS
Formula Weight 181.27
MDL Number MFCD09414738
Synonym 2-(3-Methoxyphenyl)ethanethioamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3161503

  SDS
alfa aesar™
H3161503
1g Each for $111.65

AAH3161506

  SDS
alfa aesar™
H3161506
5g Each for $373.68
Description & Specifications

Specifications

Formula Weight 181.27
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 35582-11-7
Quantity 1g
Synonym 2-(3-Methoxyphenyl)ethanethioamide
MDL Number MFCD09414738
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(3-Methoxyphenyl)thioacetamide
Melting Point 104° to 108°C
Molecular Formula C9H11NOS
Health Hazard 1 Danger
Assay 97%
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.