Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Methoxyphenyl)cyclohexanone, tech. 90%

$1,259.14 - $1,259.14

Chemical Identifiers

Chemical Name or Material 2-(3-Methoxyphenyl)cyclohexanone
CAS 15547-89-4
Purity Grade Technical Grade
Molecular Formula C13H16O2
Formula Weight 204.27
MDL Number MFCD00001630
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2261914

 
alfa aesar™
B2261914
25g Each for $1,259.14
Description & Specifications

Specifications

Formula Weight 204.27
Chemical Name or Material 2-(3-Methoxyphenyl)cyclohexanone
Purity Grade Technical Grade
Assay 90%
Molecular Formula C13H16O2
MDL Number MFCD00001630
Boiling Point 140° to 142°C (0.2mmHg)
CAS 15547-89-4
Quantity 25g
Flash Point >110°C (230°F)