missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid, 97%

$38.74 - $556.65

Chemical Identifiers

CAS 879636-95-0
Molecular Formula C12H11NO3S
Molecular Weight (g/mol) 249.284
MDL Number MFCD06857927
InChI Key WFBWIROXKGGXAW-UHFFFAOYSA-N
Synonym 2-3-methoxyphenyl-4-methyl-1,3-thiazole-5-carboxylic acid
PubChem CID 4980373
IUPAC Name 2-(3-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C2=CC(=CC=C2)OC)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54566MD View Documents Alfa Aesar™
H54566MD
250mg Each for $38.74
 
AAH5456603 View Documents Alfa Aesar™
H5456603
1g Each for $122.31
 
AAH5456606 View Documents Alfa Aesar™
H5456606
5g Each for $556.65
 
Specifications

Chemical Identifiers

879636-95-0
249.284
WFBWIROXKGGXAW-UHFFFAOYSA-N
4980373
CC1=C(SC(=N1)C2=CC(=CC=C2)OC)C(=O)O
C12H11NO3S
MFCD06857927
2-3-methoxyphenyl-4-methyl-1,3-thiazole-5-carboxylic acid
2-(3-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid

Specifications

250mg
C12H11NO3S
2-3-methoxyphenyl-4-methyl-1,3-thiazole-5-carboxylic acid
CC1=C(SC(=N1)C2=CC(=CC=C2)OC)C(=O)O
249.284
249.28
2-(3-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid
879636-95-0
MFCD06857927
WFBWIROXKGGXAW-UHFFFAOYSA-N
2-(3-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
4980373
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures