missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5456606

 View more versions of this product

Catalog No. AAH5456606



Description

Description

Specifications

Specifications

2-(3-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid
879636-95-0
MFCD06857927
WFBWIROXKGGXAW-UHFFFAOYSA-N
2-(3-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
4980373
97%
5g
C12H11NO3S
2-3-methoxyphenyl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-3-methoxyphenyl-4-methylthiazole-5-carboxylic acid, 4-methyl-2-3-methoxy _phenyl-thiazole-5-carboxylic acid, 4-methyl-2-3-methoxyphenyl thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-3-methoxyphenyl-4-methyl
CC1=C(SC(=N1)C2=CC(=CC=C2)OC)C(=O)O
249.284
249.28

Chemical Identifiers

879636-95-0
249.284
WFBWIROXKGGXAW-UHFFFAOYSA-N
4980373
CC1=C(SC(=N1)C2=CC(=CC=C2)OC)C(=O)O
C12H11NO3S
MFCD06857927
2-3-methoxyphenyl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-3-methoxyphenyl-4-methylthiazole-5-carboxylic acid, 4-methyl-2-3-methoxy _phenyl-thiazole-5-carboxylic acid, 4-methyl-2-3-methoxyphenyl thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-3-methoxyphenyl-4-methyl
2-(3-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents