missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3-Methoxyphenethyl)phenol, TCI America™

$134.45 - $456.81

Chemical Identifiers

CAS 167145-13-3
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.291
MDL Number MFCD09800498
InChI Key HGQQRAXOBYWKDV-UHFFFAOYSA-N
Synonym 2-[2-(3-Methoxyphenyl)ethyl]phenol
PubChem CID 10585448
IUPAC Name 2-[2-(3-methoxyphenyl)ethyl]phenol
SMILES COC1=CC=CC(=C1)CCC2=CC=CC=C2O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24725G View Documents TCI America
M24725G
5g Each for $456.81
 
M24721G View Documents TCI America
M24721G
1g Each for $134.45
 
Specifications

Chemical Identifiers

167145-13-3
228.291
HGQQRAXOBYWKDV-UHFFFAOYSA-N
10585448
COC1=CC=CC(=C1)CCC2=CC=CC=C2O
C15H16O2
MFCD09800498
2-[2-(3-Methoxyphenyl)ethyl]phenol
2-[2-(3-methoxyphenyl)ethyl]phenol

Specifications

White
5g
C15H16O2
2-[2-(3-Methoxyphenyl)ethyl]phenol
COC1=CC=CC(=C1)CCC2=CC=CC=C2O
228.291
228.29
≥98.0% (GC)
47°C
167145-13-3
MFCD09800498
HGQQRAXOBYWKDV-UHFFFAOYSA-N
2-[2-(3-methoxyphenyl)ethyl]phenol
10585448
Crystal-Powder at 20°C
2-(3-Methoxyphenethyl)phenol
Safety and Handling

Safety and Handling

TSCA : No