Please login to your online account to display your discounted pricing

2,3-Hexanedione +90%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC386220250

 View more versions of this product

Catalog No. AC386220250

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 2, 3-Hexanedione
Color Clear yellow
Quantity 25g
CAS 3848-24-6
CAS Min % 90.0
Health Hazard 1 Warning
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Density 0.9340g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Assay ≥90.0%
Assay Percent Range 90+%
Molecular Formula C6H10O2
Refractive Index 1.4110 to 1.4130 (20°C, 589nm)
Linear Formula CH3(CH2)2COCOCH3
Formula Weight 114.15
MDL Number MFCD00009398

Specifications

Chemical Name or Material 2, 3-Hexanedione
CAS 3848-24-6
CAS Min % 90.0
CAS Max % 100.0
Assay ≥90.0%
Assay Percent Range 90+%
Molecular Formula C6H10O2
Linear Formula CH3(CH2)2COCOCH3
Formula Weight 114.15
MDL Number MFCD00009398
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents