Please login to your online account to display your discounted pricing

2,3-Hexanedione +90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Hexanedione
CAS 3848-24-6
CAS Min % 90.0
CAS Max % 100.0
Assay ≥90.0%
Assay Percent Range 90+%
Molecular Formula C6H10O2
Linear Formula CH3(CH2)2COCOCH3
Formula Weight 114.15
MDL Number MFCD00009398
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC386220250

  SDS
thermo fisher scientific
AC386220250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥90.0%
Formula Weight 114.15
Physical Form Liquid
Quantity 25g
Color Clear yellow
Density 0.9340g/mL
CAS Max % 100.0
CAS Min % 90.0
Assay Percent Range 90+%
MDL Number MFCD00009398
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Refractive Index 1.4110 to 1.4130 (20°C, 589nm)
Packaging Glass bottle
Chemical Name or Material 2, 3-Hexanedione
Infrared Spectrum Authentic
Molecular Formula C6H10O2
Linear Formula CH3(CH2)2COCOCH3
CAS 3848-24-6
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.