Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Heptanedione, 97+%

Chemical Identifiers

Chemical Name or Material 2,3-Heptanedione
CAS 96-04-8
Molecular Formula C7H12O2
Formula Weight 128.17
MDL Number MFCD00036550
UN Number UN1224
Beilstein 1700989
Synonym Acetylvaleryl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1913606

  SDS
alfa aesar™
A1913606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1913614

  SDS
alfa aesar™
A1913614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1913622

  SDS
alfa aesar™
A1913622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.17
Chemical Name or Material 2,3-Heptanedione
TSCA TSCA
Assay ≥97%
MDL Number MFCD00036550
CAS 96-04-8
UN Number UN1224
Beilstein 1700989
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Flash Point 40°C (104°F)
Boiling Point 64°C (18mmHg)
Quantity 5g
Molecular Formula C7H12O2
Synonym Acetylvaleryl
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.