missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Fluorophenylamino)benzoic acid, 98%

Manufacturer:  Alfa Aesar™ 4385218

 View more versions of this product

Catalog No. AA4385218



Specifications

Specifications

2- (3-Fluorophenylamino)benzoic acid
158°C to 163°C
54-59-1
MFCD01675228
HYZIDPAMLUKNIR-UHFFFAOYSA-N
2-(3-fluoroanilino)benzoic acid
120158
Powder
Gray to Yellow
50g
C13H10FNO2
2-3-fluorophenyl amino benzoic acid
C1=CC=C(C(=C1)C(=O)O)NC2=CC(=CC=C2)F
231.226
231.22
98%

Chemical Identifiers

54-59-1
231.226
HYZIDPAMLUKNIR-UHFFFAOYSA-N
120158
C1=CC=C(C(=C1)C(=O)O)NC2=CC(=CC=C2)F
C13H10FNO2
MFCD01675228
2-3-fluorophenyl amino benzoic acid
2-(3-fluoroanilino)benzoic acid
Safety and Handling

Safety and Handling

RTECSNumber : CB3060000

Documents