Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Fluorophenoxy)ethylamine hydrochloride, 98%

$101.26 - $361.62

Chemical Identifiers

Chemical Name or Material 2-(3-Fluorophenoxy)ethylamine hydrochloride
CAS 120351-93-1
Molecular Formula C8H10FNO·HCl
Formula Weight 191.63
MDL Number MFCD11099553
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5168103

  SDS
alfa aesar™
H5168103
1g Each for $361.62

AAH51681MD

  SDS
alfa aesar™
H51681MD
250mg Each for $101.26
Description & Specifications

Specifications

Formula Weight 191.63
CAS 120351-93-1
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD11099553
Quantity 1g
Health Hazard 1 Warning
Chemical Name or Material 2-(3-Fluorophenoxy)ethylamine hydrochloride
Molecular Formula C8H10FNO·HCl
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.