missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Epoxynorbornane, TCI America™

Manufacturer:  TCI America E07795G

 View more versions of this product

Catalog No. E07795G



Specifications

Specifications

2,3-Epoxynorbornane
126°C
278-74-0
MFCD00798067
2,3-Epoxybicyclo[2.2.1]heptane, 3-Oxatricyclo[3.2.1.0(2,4)]octane
C1CC2CC1C3C2O3
18463
Crystal-Powder at 20°C
White-Yellow
5g
C7H10O
1325
OHNNZOOGWXZCPZ-UHFFFAOYSA-N
110.156
110.16
≥95.0% (GC)

Chemical Identifiers

278-74-0
110.156
OHNNZOOGWXZCPZ-UHFFFAOYSA-N
18463
C7H10O
MFCD00798067
2,3-Epoxybicyclo[2.2.1]heptane, 3-Oxatricyclo[3.2.1.0(2,4)]octane
C1CC2CC1C3C2O3
Safety and Handling

Safety and Handling

TSCA : No

Documents