Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Diphenylpyrazine, 98%

$45.81 - $176.30

Chemical Identifiers

Chemical Name or Material 2,3-Diphenylpyrazine
CAS 1588-89-2
Molecular Formula C16H12N2
Formula Weight 232.29
MDL Number MFCD00053000
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1470003

  SDS
alfa aesar™
A1470003
1g Each for $45.81

AAA1470006

  SDS
alfa aesar™
A1470006
5g Each for $176.30
Description & Specifications

Specifications

Formula Weight 232.29
Assay 98%
Quantity 1g
CAS 1588-89-2
Molecular Formula C16H12N2
Melting Point 119° to 122°C
Chemical Name or Material 2,3-Diphenylpyrazine
MDL Number MFCD00053000
Odor Odorless