Learn More
2,3-Diphenylmaleic anhydride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 439010250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 3-Diphenylmaleic anhydride | |
97.5 | |
Authentic | |
C16H10O3 | |
2,3-diphenylmaleic anhydride, diphenylmaleic anhydride, 3,4-diphenyl-2,5-furandione, diphenylfuran-2,5-dione, 2,3-diphenylmaleicanhydride, 2,5-furandione, 3,4-diphenyl, acmc-1asck, 2, 3,4-diphenyl, diphenyl-maleic anhydride, 2,5-furandione,3,4-diphenyl | |
C1=CC=C(C=C1)C2=C(C(=O)OC2=O)C3=CC=CC=C3 | |
250.26 | |
250.26 |
4808-48-4 | |
100.0 | |
97.5% min. (GC) | |
25g | |
OUJCFCNZIUTYBH-UHFFFAOYSA-N | |
3,4-diphenylfuran-2,5-dione | |
78530 | |
98% |
Chemical Identifiers
4808-48-4 | |
250.26 | |
2,3-diphenylmaleic anhydride, diphenylmaleic anhydride, 3,4-diphenyl-2,5-furandione, diphenylfuran-2,5-dione, 2,3-diphenylmaleicanhydride, 2,5-furandione, 3,4-diphenyl, acmc-1asck, 2, 3,4-diphenyl, diphenyl-maleic anhydride, 2,5-furandione,3,4-diphenyl | |
3,4-diphenylfuran-2,5-dione |
C16H10O3 | |
OUJCFCNZIUTYBH-UHFFFAOYSA-N | |
78530 | |
C1=CC=C(C=C1)C2=C(C(=O)OC2=O)C3=CC=CC=C3 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Co
GHS Signal Word: Warning
EINECSNumber : 225-370-2
RUO â Research Use Only