missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Diphenyl-5-(2-thienyl)tetrazolium Chloride, TCI America™

$151.25 - $685.98

Chemical Identifiers

CAS 38800-20-3
Molecular Formula C17H13ClN4S
Molecular Weight (g/mol) 340.829
MDL Number MFCD00060088
InChI Key RLHVMOZOYHDIGV-UHFFFAOYSA-M
Synonym STC
PubChem CID 6097060
IUPAC Name 2,3-diphenyl-5-thiophen-2-yltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)N2N=C(N=[N+]2C3=CC=CC=C3)C4=CC=CS4.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B0325100MG View Documents TCI America
B0325100MG
100mg Each for $151.25
 
B03251G View Documents TCI America
B03251G
1g Each for $685.98
 
Specifications

Chemical Identifiers

38800-20-3
340.829
RLHVMOZOYHDIGV-UHFFFAOYSA-M
6097060
C1=CC=C(C=C1)N2N=C(N=[N+]2C3=CC=CC=C3)C4=CC=CS4.[Cl-]
C17H13ClN4S
MFCD00060088
STC
2,3-diphenyl-5-thiophen-2-yltetrazol-2-ium;chloride

Specifications

White-Yellow
38800-20-3
MFCD00060088
RLHVMOZOYHDIGV-UHFFFAOYSA-M
2,3-diphenyl-5-thiophen-2-yltetrazol-2-ium;chloride
6097060
Crystal-Powder at 20°C
2,3-Diphenyl-5-(2-thienyl)tetrazolium Chloride
100mg
C17H13ClN4S
STC
C1=CC=C(C=C1)N2N=C(N=[N+]2C3=CC=CC=C3)C4=CC=CS4.[Cl-]
340.829
340.83
≥97.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No