missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2195403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,3-Diphenyl-2-cyclopropen-1-one | |
119°C to 121°C | |
C15H10O | |
1 g | |
14,3308 | |
HCIBTBXNLVOFER-UHFFFAOYSA-N | |
2,3-diphenylcycloprop-2-en-1-one | |
65057 | |
206.24 |
886-38-4 | |
(dry wt.), may cont up to 5% water | |
MFCD00001311 | |
608049 | |
diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec | |
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 | |
206.24 | |
CHEBI:53074 | |
99% |
Chemical Identifiers
886-38-4 | |
206.24 | |
HCIBTBXNLVOFER-UHFFFAOYSA-N | |
65057 | |
2,3-diphenylcycloprop-2-en-1-one |
C15H10O | |
MFCD00001311 | |
diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec | |
CHEBI:53074 | |
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 212-948-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only