Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethylpiperazine, cis + trans, 95%

$189.85 - $476.14

Chemical Identifiers

Chemical Name or Material 2,3-Dimethylpiperazine
CAS 84468-52-0
Name Note cis + trans
Molecular Formula C6H14N2
Formula Weight 114.19
MDL Number MFCD07373397
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2625203

  SDS
alfa aesar™
H2625203
1g Each for $476.14

AAH26252MD

  SDS
alfa aesar™
H26252MD
250mg Each for $189.85
Description & Specifications

Specifications

Formula Weight 114.19
Chemical Name or Material 2,3-Dimethylpiperazine
Name Note cis + trans
Assay 95%
Molecular Formula C6H14N2
Melting Point 81° to 90°C
MDL Number MFCD07373397
CAS 84468-52-0
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.