Please login to your online account to display your discounted pricing

2,3-Dimethylphenyl isocyanate, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Dimethylphenyl isocyanate
CAS 1591-99-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H9NO
Linear Formula (CH3)C6H3NCO
Formula Weight 147.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC366770050

  SDS
thermo fisher scientific
AC366770050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Liquid
Formula Weight 147.18
Quantity 5g
Color Clear slightly yellow
Density 1.059g/mL
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 2, 3-Dimethylphenyl isocyanate
Refractive Index 1.5380 to 1.5400 (20°C, 589nm)
Assay Percent Range 99+%
Molecular Formula C9H9NO
Linear Formula (CH3)C6H3NCO
CAS 1591-99-7
Infrared Spectrum Authentic
Recommended Storage Refrigerator +4°C
Safety and Handling

Refrigerator +4°C