Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethylphenoxyacetic acid, 98%

$74.89 - $74.89

Chemical Identifiers

Chemical Name or Material 2,3-Dimethylphenoxyacetic acid
CAS 2935-63-9
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00016814
Beilstein 2446457
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1597606

  SDS
alfa aesar™
A1597606
5g Each for $74.89
Description & Specifications

Specifications

Formula Weight 180.2
Beilstein 2446457
Assay 98%
Molecular Formula C10H12O3
CAS 2935-63-9
Quantity 5g
Chemical Name or Material 2,3-Dimethylphenoxyacetic acid
Melting Point 185° to 187°C
MDL Number MFCD00016814
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.