Please login to your online account to display your discounted pricing

(2,3-Dimethylphenoxy)acetic acid, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material (2, 3-Dimethylphenoxy)acetic acid
Assay Percent Range 98%
Molecular Formula C10H12O3
Formula Weight 180.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372470010

  SDS
thermo fisher scientific
AC372470010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC372470050

  SDS
thermo fisher scientific
AC372470050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (2, 3-Dimethylphenoxy)acetic acid
Quantity 1g
CAS 2935-63-9
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C10H12O3
Formula Weight 180.2