Please login to your online account to display your discounted pricing

2,3-Dimethylphenol 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  116521000

 View more versions of this product

Catalog No. AC116521000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Assay ≥98.5%
Formula Weight 122.17
Quantity 100g
Color Brown
Beilstein 06, 480
Merck Index 15, 10279
EINECS Number 208-395-3
TSCA TSCA
RTECS Number ZE5500000
Melting Point 72° to 74°C
CAS Max % 100.0
Chemical Name or Material 2, 3-Dimethylphenol
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H10O
Linear Formula (CH3)2C6H3OH
MDL Number MFCD00002227
CAS 526-75-0
Synonym 2, 3-Xylenol
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 2, 3-Dimethylphenol
Name Note 99%
CAS 526-75-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H10O
Linear Formula (CH3)2C6H3OH
Formula Weight 122.17
MDL Number MFCD00002227
Synonym 2, 3-Xylenol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents