Please login to your online account to display your discounted pricing

2,3-Dimethylphenol 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Dimethylphenol
Name Note 99%
CAS 526-75-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H10O
Linear Formula (CH3)2C6H3OH
Formula Weight 122.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC116520050

  SDS
thermo fisher scientific
116520050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC116525000

  SDS
thermo fisher scientific
116525000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Physical Form Crystalline Solid
Assay ≥98.5%
Quantity 5g
Color Brown
RTECS Number ZE5500000
Melting Point 72° to 74°C
TSCA TSCA
EINECS Number 208-395-3
Beilstein 06, 480
Merck Index 15, 10279
CAS Max % 100.0
Synonym 2, 3-Xylenol
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Name Note 99%
Chemical Name or Material 2, 3-Dimethylphenol
Linear Formula (CH3)2C6H3OH
MDL Number MFCD00002227
Molecular Formula C8H10O
CAS 526-75-0
Assay Percent Range 99%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.