Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethylphenol, 99%

Chemical Identifiers

Chemical Name or Material 2,3-Dimethylphenol
CAS 526-75-0
Molecular Formula C8H10O
Formula Weight 122.17
MDL Number MFCD00002227
UN Number UN2261
Beilstein 1906267
Merck Index 14,10082
Synonym 2,3-Xylenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2286022

  SDS
alfa aesar™
B2286022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2286036

  SDS
alfa aesar™
B2286036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
TSCA TSCA
Flash Point 95°C (203°F)
Melting Point 70° to 73°C
UN Number UN2261
Quantity 100g
Assay 99%
Beilstein 1906267
Health Hazard 2 GHS H Statement
H300-H310-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
RTECS Number ZE5500000
Boiling Point 218°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
Chemical Name or Material 2,3-Dimethylphenol
CAS 526-75-0
Synonym 2,3-Xylenol
Molecular Formula C8H10O
MDL Number MFCD00002227
Merck Index 14,10082
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger

Applications
2,3-Dimethylphenol is used for organic synthesis. It is used as the hardener to produce industrial furane, the raw material of producing xylenol specialized product and the assistant of the main raw material of detergent.

Solubility
Slightly soluble in water. Soluble in most solvent and aqueous alkali.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.