Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dimethylindole, 97%

$46.80 - $164.70

Chemical Identifiers

Chemical Name or Material 2,3-Dimethylindole
CAS 91-55-4
Molecular Formula C10H11N
Formula Weight 145.21
MDL Number MFCD00005617
Beilstein 116662
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1850409

  SDS
alfa aesar™
A1850409
10g Each for $46.80

AAA1850418

  SDS
alfa aesar™
A1850418
50g Each for $164.70
Description & Specifications

Specifications

Formula Weight 145.21
Chemical Name or Material 2,3-Dimethylindole
TSCA TSCA
Assay 97%
Molecular Formula C10H11N
MDL Number MFCD00005617
CAS 91-55-4
Beilstein 116662
RTECS Number NL7185000
Boiling Point 285°C (750mmHg)
Melting Point 105° to 107°C
Quantity 10g