Learn More
2,3-Dimethylbutane, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC116100010
Specifications
2, 3-Dimethylbutane | |
−29°C | |
Glass Bottle | |
0.662 | |
Colorless | |
1L | |
98.0 | |
98+% | |
(CH3)2CHCH(CH3)2 | |
2457 | |
1,1,2,2-tetramethylethane, acmc-1bmfq, biisopropyl, butane, 2,3-dimethyl, ccris 6020, ch3 2chch ch3 2, diisopropyl, dsstox_cid_5112, hsdb 76, unii-68isq7a432 | |
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N | |
2,3-dimethylbutane | |
6589 | |
Liquid |
0.6620g/mL | |
Authentic | |
1.3740 to 1.3760 | |
58.0°C | |
-129.0°C | |
79-29-8 | |
100.0 | |
C6H14 | |
MFCD00008925 | |
01,151 | |
Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents | |
CC(C)C(C)C | |
86.18 | |
86.18 | |
98+% |
Chemical Identifiers
79-29-8 | |
86.18 | |
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N | |
6589 | |
CC(C)C(C)C |
C6H14 | |
MFCD00008925 | |
1,1,2,2-tetramethylethane, acmc-1bmfq, biisopropyl, butane, 2,3-dimethyl, ccris 6020, ch3 2chch ch3 2, diisopropyl, dsstox_cid_5112, hsdb 76, unii-68isq7a432 | |
2,3-dimethylbutane |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-193-6
Promotions
-
Get a Molecular Model Set with $250 Purchase
Spend $250 or more on select chemicals and get a molecular model set at no additional cost.: 03/31/21